Assay Design



Gene Transcripts

Gene Symbol
Tbx4
Entrez Gene ID
21387
Gene Name
T-box 4
Gene Aliases
3930401C23
Chromosome Location
Chr.11: 85886343 - 85916099 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011536.2 NP_035666.2 6 - 7 807 80
NM_172798.1 NP_766386.1 5 - 6 757 80
GenBank mRNA AB221665.1 - 6 - 7 807 80
AK028407.1 - 5 - 6 691 80
AK139949.1 - 5 - 6 729 80
AK147984.1 - 5 - 6 675 80

View Full Product Details