Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278226.1 | NP_001265155.1 | 9 - 10 | 790 | 77 |
NM_010059.3 | NP_034189.1 | 11 - 12 | 955 | 77 | |
GenBank mRNA | AK076777.1 | - | 9 - 10 | 781 | 77 |
BC116767.1 | - | 11 - 12 | 840 | 77 | |
BC119081.1 | - | 11 - 12 | 840 | 77 | |
D58419.1 | - | 11 - 12 | 940 | 77 | |
D64107.1 | - | 11 - 12 | 929 | 77 |