Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278226.1 | NP_001265155.1 | 1 - 2 | 147 | 78 |
NM_010059.3 | NP_034189.1 | 1 - 2 | 147 | 78 | |
NR_103477.1 | 1 - 2 | 147 | 78 | ||
GenBank mRNA | AK076777.1 | - | 1 - 2 | 138 | 78 |
AK169570.1 | - | 1 - 2 | 147 | 78 | |
D58419.1 | - | 1 - 2 | 132 | 78 | |
D64107.1 | - | 1 - 2 | 121 | 78 |