Assay Design



Gene Transcripts

Gene Symbol
Gtpbp1
Entrez Gene ID
14904
Gene Name
GTP binding protein 1
Gene Aliases
AL022987, AW045683, C85212, GP-1, Gtpbp
Chromosome Location
Chr.15: 79690896 - 79721479 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013818.2 NP_038846.2 11 - 12 1935 97
GenBank mRNA AK004612.1 - 11 - 12 1936 97
AK150068.1 - 11 - 12 1942 97
AK150185.1 - 11 - 12 1942 97
AK158342.1 - 11 - 12 1947 97
AK170091.1 - 11 - 12 1937 97
BC037129.1 - 9 - 10 1477 97
BC046228.1 - 11 - 12 1935 97
U87965.1 - 11 - 12 1822 97

View Full Product Details