Assay Design



Gene Transcripts

Gene Symbol
Rgs21
Entrez Gene ID
624910
Gene Name
regulator of G-protein signalling 21
Gene Aliases
EG624910
Chromosome Location
Chr.1: 144519690 - 144567667 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290269.1 NP_001277198.1 1 - 2 308 88
NR_110891.1 1 - 2 308 88
GenBank mRNA BB635805.1 - 1 - 2 310 88

View Full Product Details