Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085555.1 | NP_001079024.1 | 3 - 4 | 284 | 72 |
NM_013687.3 | NP_038715.2 | 3 - 4 | 521 | 72 | |
GenBank mRNA | BC016610.1 | - | 3 - 4 | 504 | 72 |
BC058793.1 | - | 2 - 3 | 460 | 72 | |
BC089527.1 | - | 3 - 4 | 522 | 72 | |
BC089528.1 | - | 3 - 4 | 267 | 72 | |
BY098447.1 | - | 3 - 4 | 285 | 72 | |
X52127.1 | - | 1 - 2 | 72 | ||
X52128.1 | - | 3 - 4 | 499 | 72 |