Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010781.3 | NP_034911.3 | 3 - 4 | 515 | 82 |
GenBank mRNA | BC024374.1 | - | 3 - 4 | 515 | 82 |
L31853.1 | - | 3 - 4 | 509 | 82 | |
M57626.1 | - | 4 - 5 | 537 | 82 | |
X78542.1 | - | - | 509 | 82 |