Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177857.1 | NP_808525.2 | 17 - 18 | 2916 | 69 |
GenBank mRNA | AK137637.1 | - | 12 - 13 | 1523 | 69 |
AK172232.1 | - | 13 - 14 | 1638 | 69 | |
BC046440.1 | - | 13 - 14 | 1384 | 69 | |
BC060666.1 | - | 17 - 18 | 2916 | 69 |