Assay Design



Gene Transcripts

Gene Symbol
Dennd2c
Entrez Gene ID
329727
Gene Name
DENN/MADD domain containing 2C
Gene Aliases
A930010I20Rik
Chromosome Location
Chr.3: 103102654 - 103169738 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177857.1 NP_808525.2 4 - 5 1316 82
GenBank mRNA AK044389.1 - 3 - 4 1692 82
BC060666.1 - 4 - 5 1316 82

View Full Product Details