Assay Design



Gene Transcripts

Gene Symbol
Rbmx
Entrez Gene ID
19655
Gene Name
RNA binding motif protein, X chromosome
Gene Aliases
hnRNP G
Chromosome Location
Chr.X: 57383348 - 57393042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_029425.1 8 - 9 1075 71
GenBank mRNA AJ237846.1 - 8 - 9 1005 71
AK087940.1 - 2 - 3 179 71
AK165006.1 - 8 - 9 1021 71

View Full Product Details