Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146217.1 | NP_001139689.1 | 4 - 5 | 732 | 81 |
NM_022316.2 | NP_071711.2 | 4 - 5 | 732 | 81 | |
GenBank mRNA | AF070470.1 | - | 4 - 5 | 708 | 81 |
AK040931.1 | - | 4 - 5 | 733 | 81 | |
BC031804.1 | - | 4 - 5 | 712 | 81 |