Assay Design



Gene Transcripts

Gene Symbol
Ccdc78
Entrez Gene ID
381077
Gene Name
coiled-coil domain containing 78
Gene Aliases
Gm938, Gm950
Chromosome Location
Chr.17: 25785619 - 25790513 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165929.1 NP_001159401.1 5 - 6 533 77
GenBank mRNA AK140238.1 - 5 - 6 534 77

View Full Product Details