Assay Design



Gene Transcripts

Gene Symbol
Vtn
Entrez Gene ID
22370
Gene Name
vitronectin
Gene Aliases
AI256434, Vn
Chromosome Location
Chr.11: 78499120 - 78502325 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011707.2 NP_035837.1 4 - 5 892 80
GenBank mRNA AF440693.1 - 4 - 5 662 80
AK011736.1 - 4 - 5 747 80
AK090325.1 - 4 - 5 747 80
AK167494.1 - 4 - 5 745 80
BC012690.1 - 4 - 5 738 80
M77123.1 - 4 - 5 662 80
X63003.1 - 4 - 5 724 80

View Full Product Details