Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164571.1 | NP_001158043.1 | 3 - 4 | 803 | 58 |
NM_026097.3 | NP_080373.1 | 3 - 4 | 911 | 58 | |
GenBank mRNA | AF434814.1 | - | 2 - 3 | 589 | 58 |
AK006757.1 | - | 3 - 4 | 899 | 58 | |
AK007189.1 | - | 3 - 4 | 899 | 58 | |
AK015806.1 | - | 3 - 4 | 912 | 58 | |
AK028095.1 | - | 3 - 4 | 900 | 58 | |
AK169393.1 | - | 3 - 4 | 744 | 58 | |
BC118517.1 | - | 3 - 4 | 686 | 58 |