Assay Design



Gene Transcripts

Gene Symbol
Pde1b
Entrez Gene ID
18574
Gene Name
phosphodiesterase 1B, Ca2+-calmodulin dependent
Gene Aliases
Pde1b1
Chromosome Location
Chr.15: 103503034 - 103530056 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285890.1 NP_001272819.1 1 - 2 446 66
GenBank mRNA AK004772.1 - 1 - 2 53 66
CV784321.1 - 1 - 2 502 66

View Full Product Details