Assay Design



Gene Transcripts

Gene Symbol
Tulp1
Entrez Gene ID
22157
Gene Name
tubby like protein 1
Gene Aliases
Tulp1l
Chromosome Location
Chr.17: 28351515 - 28365211 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021478.1 NP_067453.1 11 - 12 1137 79
GenBank mRNA AF085681.1 - 11 - 12 1185 79
AF105711.1 - 11 - 12 1185 79
AK053880.1 - 5 - 6 640 79
AK164621.1 - 1 - 2 141 79
BC014820.1 - 11 - 12 1143 79
BC029222.1 - 11 - 12 1176 79

View Full Product Details