Assay Design
Gene Transcripts
- Gene Symbol
- Sept3
- Entrez Gene ID
-
24050
- Gene Name
- septin 3
- Gene Aliases
-
3110018K01Rik, AV154067, B530002E20Rik, Sep3
- Chromosome Location
-
Chr.15: 82272451 - 82294574 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK046722.1 |
- |
11 - 12 |
|
95 |
View Full Product Details