Assay Design



Gene Transcripts

Gene Symbol
Senp1
Entrez Gene ID
223870
Gene Name
SUMO1/sentrin specific peptidase 1
Gene Aliases
2310046A20Rik, D15Ertd528e, E330036L07Rik, suPr-2
Chromosome Location
Chr.15: 98038744 - 98093569 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144851.5 NP_659100.1 1 - 2 289 74
GenBank mRNA AK035581.1 - 1 - 2 125 74
AK054521.1 - 1 - 2 290 74
AK138013.1 - 1 - 2 90 74

View Full Product Details