Assay Design
Gene Transcripts
- Gene Symbol
- Zcrb1
- Entrez Gene ID
-
67197
- Gene Name
- zinc finger CCHC-type and RNA binding motif 1
- Gene Aliases
-
2700088M22Rik, Madp-1
- Chromosome Location
-
Chr.15: 93386106 - 93398329 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK012589.1 |
- |
4 - 5 |
323 |
132 |
View Full Product Details