Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026813.1 | NP_081089.1 | 7 - 8 | 1151 | 83 |
NM_027805.2 | NP_082081.1 | 7 - 8 | 1151 | 83 | |
GenBank mRNA | AK018403.1 | - | 7 - 8 | 1070 | 83 |
AK084684.1 | - | 7 - 8 | 1152 | 83 | |
AK173000.1 | - | 1 - 2 | 253 | 83 |