Assay Design



Gene Transcripts

Gene Symbol
Agxt2
Entrez Gene ID
268782
Gene Name
alanine-glyoxylate aminotransferase 2
Gene Aliases
AI303810, AI663818
Chromosome Location
Chr.15: 10358516 - 10410159 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031851.1 NP_001027021.1 1 - 2 107 83
GenBank mRNA AK149538.1 - 1 - 2 118 83
BC141248.1 - 1 - 2 131 83

View Full Product Details