Assay Design



Gene Transcripts

Gene Symbol
Lrrk2
Entrez Gene ID
66725
Gene Name
leucine-rich repeat kinase 2
Gene Aliases
4921513O20Rik, 9330188B09Rik, AW561911, D630001M17Rik, Gm927, cI-46
Chromosome Location
Chr.15: 91672821 - 91816124 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025730.3 NP_080006.3 3 - 4 414 70
GenBank mRNA AK052591.1 - 3 - 4 415 70
AY792512.1 - 3 - 4 357 70

View Full Product Details