Assay Design
Gene Transcripts
- Gene Symbol
- Mbtd1
- Entrez Gene ID
-
103537
- Gene Name
- mbt domain containing 1
- Gene Aliases
-
AA408199, AI194990, hemp
- Chromosome Location
-
Chr.11: 93885850 - 93946985 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC064014.1 |
- |
1 - 2 |
197 |
63 |
View Full Product Details