Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134012.3 | NP_598773.2 | 3 - 4 | 528 | 116 |
GenBank mRNA | AK141155.1 | - | 3 - 4 | 524 | 116 |
AK153919.1 | - | 5 - 6 | 363 | 116 | |
BC062907.1 | - | 3 - 4 | 528 | 116 | |
BC064014.1 | - | 5 - 6 | 632 | 116 |