Assay Design



Gene Transcripts

Gene Symbol
Rab17
Entrez Gene ID
19329
Gene Name
RAB17, member RAS oncogene family
Gene Aliases
AW413472
Chromosome Location
Chr.1: 90958133 - 90969674 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159725.2 NP_001153197.2 1 - 2 158 94
GenBank mRNA AK009707.1 - 1 - 2 159 94

View Full Product Details