Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028011.2 | NP_082287.1 | 13 - 14 | 1335 | 61 |
GenBank mRNA | AF395837.1 | - | 13 - 14 | 1299 | 61 |
AJ505007.1 | - | 13 - 14 | 1296 | 61 | |
AK009826.1 | - | 11 - 12 | 1070 | 61 | |
AK033712.1 | - | 13 - 14 | 1337 | 61 | |
AK083630.1 | - | - | 1297 | 61 | |
BC004710.1 | - | 6 - 7 | 1692 | 61 |