Assay Design



Gene Transcripts

Gene Symbol
Fam19a3
Entrez Gene ID
329731
Gene Name
family with sequence similarity 19, member A3
Gene Aliases
7530404M11Rik, TAFA3, Tafa-3
Chromosome Location
Chr.3: 104767405 - 104777547 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183224.3 NP_899047.1 1 - 2 371 95
GenBank mRNA AK078681.1 - 1 - 2 371 95
BC118531.1 - 1 - 2 61 95

View Full Product Details