Assay Design



Gene Transcripts

Gene Symbol
Mill2
Entrez Gene ID
243864
Gene Name
MHC I like leukocyte 2
Gene Aliases
Micb
Chromosome Location
Chr.7: 18839955 - 18863609 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153761.3 NP_715642.2 1 - 2 123 78
GenBank mRNA AB086267.1 - 1 - 2 123 78
AK137167.1 - 1 - 2 101 78
BC144886.1 - 1 - 2 121 78

View Full Product Details