Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023475.3 | NP_075964.1 | 7 - 8 | 653 | 94 |
GenBank mRNA | AJ245737.1 | - | - | 656 | 94 |
AJ251200.1 | - | 7 - 8 | 656 | 94 | |
AK002313.1 | - | 7 - 8 | 566 | 94 | |
AK003827.1 | - | 7 - 8 | 566 | 94 | |
AK154770.1 | - | 6 - 7 | 1861 | 94 | |
AK163336.1 | - | 1 - 2 | 499 | 94 | |
BC024048.1 | - | 7 - 8 | 534 | 94 | |
BC046342.1 | - | 7 - 8 | 538 | 94 | |
BC055431.1 | - | 7 - 8 | 665 | 94 | |
BC064069.1 | - | 5 - 6 | 703 | 94 |