Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113545.1 | NP_001107017.1 | 7 - 8 | 1121 | 62 |
NM_023063.4 | NP_075550.2 | 4 - 5 | 993 | 62 | |
GenBank mRNA | AF307845.1 | - | 7 - 8 | 1075 | 62 |
AK028186.1 | - | 4 - 5 | 934 | 62 | |
AK031698.1 | - | 4 - 5 | 937 | 62 | |
AK085065.1 | - | 4 - 5 | 987 | 62 | |
AK132314.1 | - | 4 - 5 | 939 | 62 | |
AK167220.1 | - | 4 - 5 | 553 | 62 | |
BC031490.1 | - | 7 - 8 | 1047 | 62 |