Assay Design



Gene Transcripts

Gene Symbol
Csrnp2
Entrez Gene ID
207785
Gene Name
cysteine-serine-rich nuclear protein 2
Gene Aliases
CSRNP-2, Csnrp2, taip-12
Chromosome Location
Chr.15: 100479570 - 100495239 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153407.2 NP_700456.1 3 - 4 698 65
GenBank mRNA AB091689.1 - 2 - 3 447 65
AK050864.1 - 3 - 4 719 65
AK146924.1 - 3 - 4 718 65
AK166557.1 - 3 - 4 722 65
AK202441.1 - 1 - 2 213 65
AK203108.1 - 1 - 2 180 65
BC035295.1 - 3 - 4 698 65

View Full Product Details