Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083901.1 | NP_001077370.1 | 6 - 7 | 783 | 77 |
NM_133680.3 | NP_598441.3 | 5 - 6 | 880 | 77 | |
GenBank mRNA | AK146596.1 | - | 6 - 7 | 776 | 77 |
BC033469.1 | - | 5 - 6 | 842 | 77 | |
CN709077.1 | - | 2 - 3 | 209 | 77 |