Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122963.1 | NP_001116435.1 | 8 - 9 | 2133 | 78 |
NM_028487.4 | NP_082763.3 | 9 - 10 | 2193 | 78 | |
GenBank mRNA | AK082704.1 | - | 8 - 9 | 1951 | 78 |
AK135728.1 | - | 9 - 10 | 2124 | 78 | |
AK136894.1 | - | 9 - 10 | 1972 | 78 | |
AK145318.1 | - | 8 - 9 | 2122 | 78 | |
AK151283.1 | - | 7 - 8 | 1354 | 78 | |
AK163370.1 | - | 9 - 10 | 2041 | 78 | |
AK163390.1 | - | 9 - 10 | 2195 | 78 | |
AK169406.1 | - | 9 - 10 | 1988 | 78 | |
AY382529.1 | - | 9 - 10 | 1991 | 78 | |
BC016083.1 | - | 7 - 8 | 987 | 78 | |
BC067075.1 | - | 8 - 9 | 1907 | 78 | |
BC076635.1 | - | 8 - 9 | 1963 | 78 | |
BC094888.1 | - | 8 - 9 | 1926 | 78 | |
BC096442.1 | - | 9 - 10 | 2008 | 78 |