Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026075.2 | NP_080351.2 | 2 - 3 | 181 | 74 |
GenBank mRNA | AK013429.1 | - | 2 - 3 | 176 | 74 |
AK013737.1 | - | 2 - 3 | 195 | 74 | |
AK014102.1 | - | 2 - 3 | 188 | 74 | |
AK018357.1 | - | 2 - 3 | 206 | 74 | |
AK020113.1 | - | 2 - 3 | 202 | 74 | |
AK030777.1 | - | - | 2039 | 74 | |
AK135887.1 | - | - | 227 | 74 | |
BC027133.1 | - | 2 - 3 | 159 | 74 | |
BC038302.1 | - | 2 - 3 | 214 | 74 | |
BC096661.1 | - | 2 - 3 | 181 | 74 | |
BC116993.1 | - | 2 - 3 | 152 | 74 | |
BC116999.1 | - | 2 - 3 | 152 | 74 |