Assay Design



Gene Transcripts

Gene Symbol
Krt15
Entrez Gene ID
16665
Gene Name
keratin 15
Gene Aliases
AI528832, K15, Krt1-15
Chromosome Location
Chr.11: 100131759 - 100135949 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008469.2 NP_032495.2 4 - 5 977 81
GenBank mRNA BC057934.1 - 4 - 5 956 81
CN456368.1 - 4 - 5 612 81

View Full Product Details