Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026191.2 | NP_080467.3 | 4 - 5 | 657 | 128 |
GenBank mRNA | AK004610.1 | - | 4 - 5 | 621 | 128 |
AK010512.1 | - | 4 - 5 | 637 | 128 | |
AK029935.1 | - | 4 - 5 | 630 | 128 | |
AK031000.1 | - | 4 - 5 | 604 | 128 | |
AK135592.1 | - | 4 - 5 | 593 | 128 | |
AK216252.1 | - | 1 - 2 | 43 | 128 | |
BC058276.1 | - | 4 - 5 | 575 | 128 |