Assay Design



Gene Transcripts

Gene Symbol
Cyp2d26
Entrez Gene ID
76279
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 26
Gene Aliases
1300006E06Rik
Chromosome Location
Chr.15: 82790107 - 82794452 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029562.2 NP_083838.1 2 - 3 439 79
GenBank mRNA AK004915.1 - 2 - 3 440 79
BC023241.1 - 2 - 3 428 79

View Full Product Details