Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183017.2 | NP_898838.2 | 1 - 2 | 208 | 68 |
GenBank mRNA | AK084879.1 | - | 1 - 2 | 171 | 68 |
AK134345.1 | - | 1 - 2 | 174 | 68 | |
AK150116.1 | - | 1 - 2 | 174 | 68 | |
AM690756.1 | - | 1 - 2 | 161 | 68 |