Assay Design



Gene Transcripts

Gene Symbol
Osmr
Entrez Gene ID
18414
Gene Name
oncostatin M receptor
Gene Aliases
OSMRB
Chromosome Location
Chr.15: 6813568 - 6875246 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011019.3 NP_035149.2 11 - 12 1703 100
GenBank mRNA AF058805.1 - 11 - 12 1673 100
AK157508.1 - 6 - 7 876 100
BC137703.1 - 10 - 11 1601 100

View Full Product Details