Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178919.4 | NP_849250.1 | 13 - 14 | 2017 | 94 |
GenBank mRNA | AK042427.1 | - | 13 - 14 | 1979 | 94 |
AK084606.1 | - | 13 - 14 | 1843 | 94 | |
AK149916.1 | - | 13 - 14 | 1969 | 94 | |
AK149934.1 | - | 13 - 14 | 2018 | 94 | |
AK158371.1 | - | 12 - 13 | 2073 | 94 | |
AK171488.1 | - | 13 - 14 | 1998 | 94 | |
BC026448.1 | - | 12 - 13 | 1797 | 94 | |
BC038039.1 | - | 12 - 13 | 2519 | 94 | |
BC058973.1 | - | 9 - 10 | 2299 | 94 | |
BC079532.1 | - | 13 - 14 | 1913 | 94 |