Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013654.3 | NP_038682.1 | 2 - 3 | 257 | 78 |
GenBank mRNA | AF128193.1 | - | 2 - 3 | 222 | 78 |
AF128194.1 | - | 2 - 3 | 222 | 78 | |
AK078827.1 | - | 2 - 3 | 258 | 78 | |
BC061126.1 | - | 2 - 3 | 255 | 78 | |
BY163435.1 | - | 2 - 3 | 259 | 78 | |
L04694.1 | - | 2 - 3 | 219 | 78 | |
S50588.1 | - | 2 - 3 | 205 | 78 | |
S71251.1 | - | 2 - 3 | 237 | 78 | |
Z12297.1 | - | 2 - 3 | 205 | 78 |