Assay Design



Gene Transcripts

Gene Symbol
Ampd1
Entrez Gene ID
229665
Gene Name
adenosine monophosphate deaminase 1
Gene Aliases
AI553520, Ampd-1
Chromosome Location
Chr.3: 103073568 - 103099721 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033303.2 NP_001028475.2 4 - 5 457 78
GenBank mRNA AK132524.1 - 4 - 5 448 78
AK137549.1 - 4 - 5 449 78

View Full Product Details