Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008979.2 | NP_033005.1 | 10 - 11 | 956 | 78 |
NR_104070.1 | 12 - 13 | 981 | 78 | ||
GenBank mRNA | AK153812.1 | - | 12 - 13 | 982 | 78 |
BC055377.1 | - | 10 - 11 | 878 | 78 | |
M90388.1 | - | 10 - 11 | 941 | 78 |