Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163552.1 | NP_001157024.1 | 6 - 7 | 1324 | 94 |
NM_026193.2 | NP_080469.2 | 5 - 6 | 1537 | 94 | |
GenBank mRNA | AK007733.1 | - | 5 - 6 | 1426 | 94 |
BC054092.1 | - | 6 - 7 | 1335 | 94 | |
BC056200.2 | - | 5 - 6 | 1425 | 94 |