Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163552.1 | NP_001157024.1 | 9 - 10 | 1725 | 57 |
NM_001163553.1 | NP_001157025.1 | 7 - 8 | 1854 | 57 | |
NM_026193.2 | NP_080469.2 | 8 - 9 | 1938 | 57 | |
GenBank mRNA | AK007733.1 | - | 8 - 9 | 1827 | 57 |
AK165570.1 | - | 7 - 8 | 1774 | 57 | |
BC054092.1 | - | 9 - 10 | 1736 | 57 | |
BC056200.2 | - | 8 - 9 | 1826 | 57 |