Assay Design



Gene Transcripts

Gene Symbol
Mks1
Entrez Gene ID
380718
Gene Name
Meckel syndrome, type 1
Gene Aliases
AK190930, B8d3
Chromosome Location
Chr.11: 87852928 - 87863679 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039684.2 NP_001034773.2 6 - 7 665 90
GenBank mRNA AK030982.1 - 6 - 7 645 90
AK138267.1 - 5 - 6 635 90
AK170482.1 - 6 - 7 664 90
BC150995.1 - 6 - 7 733 90
BC158122.1 - 6 - 7 825 90
DQ177342.1 - 6 - 7 665 90

View Full Product Details