Assay Design



Gene Transcripts

Gene Symbol
Mmp25
Entrez Gene ID
240047
Gene Name
matrix metallopeptidase 25
Gene Aliases
F730048C11Rik, Leukolysin, MMP20, MT6-MMP
Chromosome Location
Chr.17: 23629458 - 23645269 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033339.3 NP_001028511.1 8 - 9 1740 70
GenBank mRNA AK089273.1 - 8 - 9 1479 70
AK089533.1 - 8 - 9 1479 70
AK137491.1 - 8 - 9 1342 70
AK154458.1 - 8 - 9 1742 70
AK171493.1 - 8 - 9 1479 70
BC112379.1 - 8 - 9 1426 70

View Full Product Details