Assay Design



Gene Transcripts

Gene Symbol
Mmp25
Entrez Gene ID
240047
Gene Name
matrix metallopeptidase 25
Gene Aliases
F730048C11Rik, Leukolysin, MMP20, MT6-MMP
Chromosome Location
Chr.17: 23629458 - 23645269 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033339.3 NP_001028511.1 9 - 10 1990 64
GenBank mRNA AK089273.1 - 9 - 10 1730 64
AK089533.1 - 9 - 10 1729 64
AK154458.1 - 9 - 10 1992 64
AK171493.1 - 9 - 10 1729 64
BC059059.1 - 7 - 8 2739 64
BC112379.1 - 9 - 10 1676 64

View Full Product Details