Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178594.3 | NP_848709.2 | 3 - 4 | 542 | 94 |
GenBank mRNA | AK143276.1 | - | 3 - 4 | 543 | 94 |
AK145664.1 | - | 3 - 4 | 542 | 94 | |
AY280973.1 | - | 3 - 4 | 442 | 94 | |
AY322147.1 | - | 3 - 4 | 442 | 94 | |
AY346099.1 | - | 3 - 4 | 522 | 94 |