Assay Design



Gene Transcripts

Gene Symbol
Dpt
Entrez Gene ID
56429
Gene Name
dermatopontin
Gene Aliases
1810032B19Rik, 5033416F05Rik, EQ-1, Eq1
Chromosome Location
Chr.1: 164796732 - 164824266 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019759.2 NP_062733.1 1 - 2 306 71
GenBank mRNA AF143374.1 - 1 - 2 306 71
AK019890.1 - 1 - 2 395 71
BC046420.1 - 1 - 2 332 71

View Full Product Details